Learn More
Sino Biological Human respiratory syncytial virus (RSV) Fusion glycoprotein / RSV-F Antibody 100 µL
Catalog No. 5000750316
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Catalog No. 50-007-50316 Supplier Sino Biological Supplier No. 11049D001100
This product is a recombinant monoclonal antibody expressed from HEK293 cells.