Learn More
Sino Biological Human respiratory syncytial virus (RSV) glycoprotein G / RSV-G Antibody 100 µL
Catalog No. 5000749943
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Catalog No. 50-007-49943 Supplier Sino Biological Supplier No. 40830D001100
This product is a recombinant monoclonal antibody expressed from HEK293 cells.