Learn More
Raybiotech Inc Human Ret ELISA Kit
Supplier:  Raybiotech Inc ELHRET1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human Ret. Other species not yet determined.
Catalog No. 50-194-9685