Learn More
Biomatik Corporation Human Retinaldehyde Binding Protein 1 (RLBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU08445
Encompass_Preferred
Human Retinaldehyde Binding Protein 1 (RLBP1) ELISA Kit for detecting RLBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.54ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12271. Gene ID: 6017. Target Synonyms: CRALBP; Cellular retinaldehyde-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4667