Learn More
Biomatik Corporation Human Retinoic Acid Receptor Beta (RARb) ELISA Kit
Supplier:  Biomatik Corporation EKU07095
Encompass_Preferred
Human Retinoic Acid Receptor Beta (RARb) ELISA Kit for detecting RARb in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10826. Gene ID: 5915. Target Synonyms: HAP; NR1B2; RRB2; RAR-epsilon; HBV-activated protein; Nuclear receptor subfamily 1 group B member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5508