Learn More
Biomatik Corporation Human Retinol Binding Protein 1, Cellular (RBP1) CLIA Kit
Supplier:  Biomatik Corporation EKU08869
Encompass_Preferred
Human Retinol Binding Protein 1, Cellular (RBP1) CLIA Kit for detecting RBP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.12ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P09455. Gene ID: 5947. Target Synonyms: RBP-1; CRABP-I; CRBP; CRBP1; RBPC; Cellular retinol-binding protein I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6040