Learn More
Sino Biological Anti-Resistin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13055MM19100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Resistin (rh Resistin; NP_065148.1; Ser17-Pro108). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4334