Learn More
Sino Biological Anti-RFXANK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 202816T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human RFXANK fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8160