Learn More
BIOHIPPO Human RGS19 Microsample
SDP
Catalog No. 5000314120
Encompass
Human RGS19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RGS19 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human RGS19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human RGS19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human RGS19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14120 Supplier BIOHIPPO Supplier No. BHE15210853
Human RGS19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RGS19 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human RGS19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human RGS19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human RGS19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color