Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RGS2 (aa 121-198) Control Fragment Recombinant Protein

Catalog No. RP104559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104559 Supplier Invitrogen™ Supplier No. RP104559
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Regulator of G protein signaling (RGS) family members are regulatory molecules that act as GTPase activating proteins (GAPs) for G alpha subunits of heterotrimeric G proteins. RGS proteins are able to deactivate G protein subunits of the Gi alpha, Go alpha and Gq alpha subtypes. They drive G proteins into their inactive GDP-bound forms. Regulator of G protein signaling 2 belongs to this family. The protein acts as a mediator of myeloid differentiation and may play a role in leukemogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41220
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5997
Name Human RGS2 (aa 121-198) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias cell growth-inhibiting gene 31 protein; cell growth-inhibiting protein 31; G0 to G1 switch regulatory 8, 24kD; G0/G1 switch regulatory protein 8; G0S8; GIG31; GOS8; regulator of G protein signaling 2; regulator of G protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2; regulator of G-protein signaling 2, 24kDa; regulator of G-protein signaling protein 2; regulator of G-protein signalling 2, 24kDa; Rgs2
Common Name RGS2
Gene Symbol RGS2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KTKSPQKLSSKARKIYTDFIEKEAPKEINIDFQTKTLIAQNIQEATSGCFTTAQKRVYSLMENNSYPRFLESEFYQDL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.