Learn More
Sino Biological Anti-RHEBL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101739T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human RHEBL1, and purified by antigen affinity chromatography.
Catalog No. 50-241-1585