Learn More
Sino Biological Recombinant Human, Rhesus ERBB4/Her4 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10363H08H1
Encompass_Preferred
A DNA sequence encoding the human / rhesus ERBB4 (NP_005226.1) (Met1-Arg649) with a C-terminal polyhistidine tag was expressed. Human and Rhesus ERBB4 sequences are identical.
Catalog No. 50-224-6826