Learn More
ATCC Human rhinovirus 17; Strain: 33342
Catalog No. 502384287
Encompass
Human rhinovirus 17 strain 33342 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was presumed to be isolated from a throat swab from an adult with an upper respiratory illness in 1959 and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.
Catalog No. 50-238-4287 Supplier ATCC Supplier No. VR1663
Human rhinovirus 17 strain 33342 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was presumed to be isolated from a throat swab from an adult with an upper respiratory illness in 1959 and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.