Learn More
ATCC Human rhinovirus 1A; Strain: 2060
Catalog No. 502384022
Encompass
Human rhinovirus 1A strain 2060 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated from the naso-pharyngeal washings of a patient with mild respiratory illness in Ohio and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.
Catalog No. 50-238-4022 Supplier ATCC Supplier No. VR1559
Human rhinovirus 1A strain 2060 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated from the naso-pharyngeal washings of a patient with mild respiratory illness in Ohio and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in respiratory disease research.