Learn More
ATCC Human Rhinovirus 3; Strain: FEB
Supplier:  ATCC VR483
Encompass
Rhinovirus 3 strain FEB is propagated in HeLa cells (ATCC CCL-2). This strain was isolated from the throat washings of a human patient. The product has applications in respiratory disease research.
Catalog No. 50-238-2419
 Restricted Item