Learn More
ATCC Human rhinovirus 39; Strain: 209
Supplier:  ATCC VR340
Encompass
Human rhinovirus 39 strain 209 is propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated from the nasal washing of an adult female in North Carolina and was deposited by the National Institute of Allergy and Infectious Diseases. This product has applications in respiratory disease research.
Catalog No. 50-238-2873