Learn More
Creative Biomart Human Rho(D) immune globulin
Catalog No. 502084125
Encompass_Preferred
Rho (D) immune globulin is available as a sterile, lyophilized or liquid gamma globulin (IgG) fraction containing antibodies to the Rh0 (D) antigen (D antigen) under the name Rhophylac (IM/IV). Immune globullin was purified via ion-exchange chromatography method and prepared from pools of human plasma, where the donors are Rho (D)-negative donors who have been immunized with Rho(D)-positive RBCs.
Catalog No. 50-208-4125 Supplier Creative Biomart Supplier No. THP0074
Rho (D) immune globulin is available as a sterile, lyophilized or liquid gamma globulin (IgG) fraction containing antibodies to the Rh0 (D) antigen (D antigen) under the name Rhophylac (IM/IV). Immune globullin was purified via ion-exchange chromatography method and prepared from pools of human plasma, where the donors are Rho (D)-negative donors who have been immunized with Rho(D)-positive RBCs.