Learn More
Sino Biological Recombinant Anti-RhoA Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 12441R036100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human RhoA (rh RhoA; Catalog#12441-H07B; P61586-1; Met1-Leu193).
Catalog No. 50-241-2431