Learn More
Sino Biological Anti-RhoA Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 12441MM03P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RhoA (rh RhoA; Catalog#12441-H07B; P61586-1; Met 1-Leu 193) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8519