Learn More
Sino Biological Human RHOA MAb 100ul (Sino Biological)
Supplier:  Sino Biological 12441R103100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human RhoA (rh RhoA; Catalog#12441-H07B; P61586-1; Met1-Leu193).
Catalog No. 50-237-3657