Learn More
American Research Products Inc Human Ribonuclease 7 RNASE7
Catalog No. 5000327616
Encompass
Human Ribonuclease 7 (RNASE7) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma urine saliva cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27616 Supplier American Research Products Inc Supplier No. CSBEL019798HU
Human Ribonuclease 7 (RNASE7) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma urine saliva cell culture supernates tissue homogenates cell lysates Method Sandwich