Learn More
Biomatik Corporation Human Ribonuclease A7 (RNASE7) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12210
Encompass_Preferred
Human Ribonuclease A7 (RNASE7) High-sensitive ELISA Kit for detecting RNASE7 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H1E1. Gene ID: 84659. Target Synonyms: SAP-2; Rnase-A7; Ribonuclease,RNase A Family 7; Skin-derived antimicrobial protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5957