Learn More
Biomatik Corporation Human Ribonuclease III, Nuclear (RNASEN) ELISA Kit
Supplier:  Biomatik Corporation EKU07126
Encompass_Preferred
Human Ribonuclease III, Nuclear (RNASEN) ELISA Kit for detecting RNASEN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NRR4. Gene ID: 29102. Target Synonyms: RnaseIII; RNASE3L; Rnase-III; Etohi2; DROSHA; RN3; RNASE3-L; Ribonuclease Type III; Double-Stranded RNA-Specific Endoribonuclease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6499