Learn More
Biomatik Corporation Human Ribophorin I (RPN1) ELISA Kit
Supplier:  Biomatik Corporation EKU07133
Encompass_Preferred
Human Ribophorin I (RPN1) ELISA Kit for detecting RPN1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04843. Gene ID: 6184. Target Synonyms: RPNI; OST1; RBPH1; Dolichyl-Diphosphooligosaccharide-Protein Glycosyltransferase Subunit 1; Dolichyl-diphosphooligosaccharide protein glycosyltransferase 67 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6498