Learn More
Biomatik Corporation Human Ribosomal Protein L13A (RPL13A) ELISA Kit
Supplier:  Biomatik Corporation EKU07134
Encompass_Preferred
Human Ribosomal Protein L13A (RPL13A) ELISA Kit for detecting RPL13A in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40429. Gene ID: 23521. Target Synonyms: TSTA1; Tissue Specific Transplantation Antigen 1; 60S ribosomal protein L13a; 23 kDa highly basic protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6889