Learn More
Biomatik Corporation Human Ribosomal Protein S19 (RPS19) ELISA Kit
Supplier:  Biomatik Corporation EKU11407
Encompass_Preferred
Human Ribosomal Protein S19 (RPS19) ELISA Kit for detecting RPS19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39019. Gene ID: 6223. Target Synonyms: DBA; Diamond-Blackfan Anemia; 40S ribosomal protein S19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4783