Learn More
Biomatik Corporation Human Ribosomal Protein S6 Kinase Alpha 1 (RPS6Ka1) ELISA Kit
Supplier:  Biomatik Corporation EKU07137
Encompass_Preferred
Human Ribosomal Protein S6 Kinase Alpha 1 (RPS6Ka1) ELISA Kit for detecting RPS6Ka1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15418. Gene ID: 6195. Target Synonyms: HU-1; MAPKAPK1A; RSK; RSK1; S6K-alpha 1; 90 kDa ribosomal protein S6 kinase 1; MAP kinase-activated protein kinase 1a; Ribosomal S6 kinase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6260