Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RIM2 Control Fragment Recombinant Protein

Catalog No. RP107501
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107501 Supplier Invitrogen™ Supplier No. RP107501
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84724. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UQ26
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 9699
Name Human RIM2 Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2810036I15Rik; AW048769; KIAA0751; mKIAA0751; Nim2; non-small cell lung cancer RimL3a protein; non-small cell lung cancer RimL3c protein; nuclear protein; OBOE; Rab3 interacting protein 1; Rab3 interacting protein 2; RAB3 interacting protein 3; Rab-3-interacting molecule 2; rab3-interacting molecule 2; Rab3-interacting protein; rab-3-interacting protein 2; rab3-interacting protein 2; Rab-3-interacting protein 3; Rab3ip2; RAB3IP3; regulating synaptic membrane exocytosis 2; regulating synaptic membrane exocytosis protein 2; RIM 2; Rim2; RIM2 (CT); Rim2(+40A); Rim2(+44A); Rim2(+4A); Rims2; Serg2; synaptic exocytosis regulator 2; synaptotagmin 3, related sequence; Syt3-rs
Common Name RIM2
Gene Symbol Rims2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MDIEERNRQMKINKYKQVAGSDPRLEQDYHSKYRSGWDPHRGADNVSTKSS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.