Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RIM2 Synthetic Peptide

Catalog No. PEP1600 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1600 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1600 Supplier Invitrogen™ Supplier No. PEP1600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acids near the carboxy terminus of human RIM2. PEP-1600 can be used as a blocking peptide with polyclonal antibody PA5-34557. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.

Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. Rim1 and Rim2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like Rim1, Rim2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9699
Species Human
Purification Method purified
Accession Number Q9UQ26
Common Name RIM2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2810036I15Rik; AW048769; KIAA0751; mKIAA0751; Nim2; non-small cell lung cancer RimL3a protein; non-small cell lung cancer RimL3c protein; nuclear protein; OBOE; Rab3 interacting protein 1; Rab3 interacting protein 2; RAB3 interacting protein 3; Rab-3-interacting molecule 2; rab3-interacting molecule 2; Rab3-interacting protein; rab-3-interacting protein 2; rab3-interacting protein 2; Rab-3-interacting protein 3; Rab3ip2; RAB3IP3; regulating synaptic membrane exocytosis 2; regulating synaptic membrane exocytosis protein 2; RIM 2; Rim2; RIM2 (CT); Rim2(+40A); Rim2(+44A); Rim2(+4A); Rims2; Serg2; synaptic exocytosis regulator 2; synaptotagmin 3, related sequence; Syt3-rs
Gene Symbol Rims2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.