Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RIN3 Partial ORF (NP_079108, 328 a.a. - 426 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89960045 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-960-045 10 μg
89-960-046 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-960-045 Supplier Abnova™ Supplier No. H00079890Q01S

Used for AP, Array, ELISA, WB-Re

RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (see RAB5A; MIM 179512) (Kajiho et al., 2003 [PubMed 12972505]).[supplied by OMIM]

Sequence: GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTP

Specifications

Accession Number NP_079108
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 79890
Molecular Weight (g/mol) 36.63kDa
Name RIN3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen GPPDHPNQPPMMTCERLPCPTAGLGPLREEAMKPGAASSPLQQVPAPPLPAKKNLPTAPPRRRVSERVSLEDQSPGMAAEGDQLSLPPQGTSDGPEDTP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp762H1613/FLJ11700/FLJ22439
Common Name RIN3
Gene Symbol RIN3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.