Learn More
Biomatik Corporation Human Ring Finger Protein 39 (RNF39) ELISA Kit
Supplier:  Biomatik Corporation EKU07139
Encompass_Preferred
Human Ring Finger Protein 39 (RNF39) ELISA Kit for detecting RNF39 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H2S5. Gene ID: 80352. Target Synonyms: HZF; HZFW; HZFw1; LIRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5000