Learn More
BIOHIPPO Human RLN3 Microsample
SDP
Catalog No. 5000313941
Encompass
Human RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 44 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13941 Supplier BIOHIPPO Supplier No. BHE15210553
Human RLN3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of RLN3 in Serum plasma and other biological fluids It provides a detection range of 156 25-10000 pg/mL with a sensitivity of 44 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human RLN3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human RLN3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human RLN3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color