Learn More
Sino Biological Anti-Ribonuclease A Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13468MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RNase A / Ribonuclease A / RNASE1 (rh RNase A / Ribonuclease A / RNASE1; Catalog#13468-H08H; P07998; Met1-Thr156). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-255-4236