Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RNF168 Synthetic Peptide

Catalog No. PEP0852 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0852 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0852 Supplier Invitrogen™ Supplier No. PEP0852
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 18 amino acids near the carboxy terminus of human RNF168. PEP-0852 can be used as a blocking peptide with polyclonal antibody PA5-20738.

RNF168 was identified as a chromatin-associated RING finger protein that acts as a ubiquitin ligase both in vitro and in vivo. RNF168 targets histones H2A and H2AX, but not H2B, forming K63 polyubiquitin chains. Upon formation of DNA double strand breaks, RNF168 is recruited to the site of DNA damage where it co-localizes with -gammaH2AX and 53BP1 in an RNF8-dependent manner. This localization of RNF168 increases the local concentration of ubiquinated proteins to the threshold required for retention of the proteins 53BP1 and BRCA1, facilitating the downstream signaling cascade. Thus, RNF168 defines a new pathway demonstrating a functional cooperation between E3 ligases in genome maintenance. At least three isoforms of RNF168 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 165918
Species Human
Purification Method purified
Accession Number Q8IYW5
Common Name RNF168
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 3110001H15Rik; E3 ubiquitin-protein ligase RNF168; FLJ35794; hRNF168; RING finger protein 168; ring finger protein 168, E3 ubiquitin protein ligase; ring fnger protein 168; RING-type E3 ubiquitin transferase RNF168; RNF168
Gene Symbol RNF168
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.