Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RNF186 (aa 75-148) Control Fragment Recombinant Protein

Catalog No. RP95715
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP95715 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP95715 Supplier Invitrogen™ Supplier No. RP95715
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56972. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The RNF186 gene encodes the RING finger protein 186, a member of the RING-type E3 ubiquitin ligase family. RNF186 is involved in the ubiquitin-proteasome pathway, playing a crucial role in tagging target proteins for degradation, thus influencing protein turnover and cellular homeostasis. This process is essential for regulating various cellular functions, including signal transduction, apoptosis, and response to stress. RNF186 has been implicated in the unfolded protein response (UPR) within the endoplasmic reticulum, where it helps in managing protein folding and maintaining cellular homeostasis, particularly under conditions of endoplasmic reticulum stress. Alterations in RNF186 expression or function have been associated with disorders such as inflammatory bowel disease and ulcerative colitis, suggesting its role in gastrointestinal health and disease. Research into RNF186 focuses on its regulatory mechanisms within the ubiquitin-proteasome system and its potential as a therapeutic target for conditions associated with protein misfolding and cellular stress responses.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NXI6
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 54546
Name Human RNF186 (aa 75-148) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 9130020G10Rik; E3 ubiquitin-protein ligase RNF186; RING finger protein 186; RNF186
Common Name RNF186
Gene Symbol RNF186
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DNTWSITCPLCRKVTAVPGGLICSLRDHEAVVGQLAQPCTEVSLCPQGLVDPADLAAGHPSLVGEDGQDEVSAN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.