Learn More
BIORTUS USA INC Human RNF4, biotinylated Protein, 100UG
Catalog No. NC2953305
Encompass_Preferred
Gene Name: RNF4. Uniprot: P78317. Construct:biotinylated-GG-RNF4(G120-I190 end). Expression Host: E. coli. Purity: 90%.
Catalog No. NC2953305 Supplier BIORTUS USA INC Supplier No. BP1171400B
Gene Name: RNF4. Uniprot: P78317. Construct:biotinylated-GG-RNF4(G120-I190 end). Expression Host: E. coli. Purity: 90%.