Learn More
Bioworld HUMAN RO-52/SS-A (RECOMBINANT)
Supplier:  Bioworld 220604711
Encompass_Preferred
Recombinant Human RO-52/SS-A Source: E.Coli Background: The SS-A/Ro particle contains a hY RNA (human cytoplasmic RNA) and associated proteins: a 60 kDa and a 52 kDa protein. The 52 kDa protein is not directly bound to the hY RNA but...
Catalog No. 50-253-567