Learn More
ACROBiosystems Biotinylated Human ROBO1 Protein, His,Avitag™ (MALS verified)
Catalog No. 502718984
Encompass_Preferred
ROBO1 is a member of the ROBO immunoglobulin superfamily of proteins, and it plays a crucial role in cell motility and migration during embryogenesis and organogenesis. In addition, evidence showed that ROBO1 might drive migration and invasion in malignant cells, such as glioma and breast cancer, which might play a role in cancer aggressiveness. In contrast, some studies suggested that ROBO1 pathways play a key role in tumors by acting as a tumor suppressor, especially in cell invasion.
Catalog No. 50-271-8984 Supplier ACROBiosystems Supplier No. RB1H82E5200UG
ROBO1 is a member of the ROBO immunoglobulin superfamily of proteins, and it plays a crucial role in cell motility and migration during embryogenesis and organogenesis. In addition, evidence showed that ROBO1 might drive migration and invasion in malignant cells, such as glioma and breast cancer, which might play a role in cancer aggressiveness. In contrast, some studies suggested that ROBO1 pathways play a key role in tumors by acting as a tumor suppressor, especially in cell invasion.