Learn More
Sino Biological Human ROBO1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 30073H02H100
Encompass_Preferred
A DNA sequence encoding the human ROBO1 (Q9Y6N7-1) (Met1-Pro897) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1698