Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ROBO4 ELISA Kit

Catalog No. EH394RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH394RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH394RBX10 Supplier Invitrogen™ Supplier No. EH394RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human ROBO4 quantitates human ROBO4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ROBO4.

Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins. Involved in the maintenance of endothelial barrier organization and function (PubMed:30455415). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WZ75
Assay Range 0.61-150 ng/mL
Assay Sensitivity 0.61 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO36741200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO3674;
Gene ID (Entrez) 54538
Gene Symbol ROBO4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ROBO4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.