Learn More
Sino Biological Human ROBO4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11960RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ROBO4 (rh ROBO4; Catalog#11960-H08H; Q8WZ75-1; Met 1-Glu 469). ROBO4 specific IgG was purified by Human ROBO4 affinity chromatography.
Catalog No. 50-237-0770