Learn More
Sino Biological Recombinant Human ROR1 Protein, Amine-Labeled Biotinylated, HPLC-verified
Supplier:  Sino Biological 13968HCCH1B100
Encompass_Preferred
A DNA sequence encoding the Human ROR1 (NP_005003.2) (Met1-Glu403) was expressed with six amino acids (LEVLFQ) at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4151