Learn More
Sino Biological Human ROR1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 50623T48100
Encompass_Preferred
A DNA sequence encoding the Human ROR1 (NP_005003.2)(Met1-Glu403) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-4096