Learn More
ACROBiosystems Human ROR1 Protein, Mouse IgG2a Fc Tag
Catalog No. 502055266
Encompass_Preferred
Recombinant Protein;1MG;Human ROR1, Mouse IgG2a Fc Tag (RO1-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.9 kDa.
Catalog No. 50-205-5266 Supplier ACROBiosystems Supplier No. RO1H52561MG
Recombinant Protein;1MG;Human ROR1, Mouse IgG2a Fc Tag (RO1-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 68.9 kDa.