Learn More
ATCC Human rotavirus; Strain: WA (TC-adapted); Quantitative RNA
Catalog No. 502384526
Encompass
Quantitative Genomic RNA from Rotavirus A strain Wa (TC adapted) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-4526 Supplier ATCC Supplier No. VR2018DQ
Quantitative Genomic RNA from Rotavirus A strain Wa (TC adapted) can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.