Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RPAIN Synthetic Peptide

Catalog No. PEP0446 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0446 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0446 Supplier Invitrogen™ Supplier No. PEP0446
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 17 amino acids near the carboxy terminus of human RPA interacting protein PEP-0446 can be used as a blocking peptide with polyclonal antibody PA5-20326.

Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 84268
Species Human
Purification Method purified
Accession Number Q86UA6
Common Name RPAIN
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2400006N03Rik; hRIP; nuclear transporter; RAP interaction protein; RAP interaction protein alpha; RGD1308492; RIP; RPA interacting protein; RPA interacting protein delta 2; RPA interacting protein epsilon; RPAIN; RPA-interacting protein
Gene Symbol RPAIN
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.