Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RPAP1 (aa 101-215) Control Fragment Recombinant Protein

Catalog No. RP98046
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98046 Supplier Invitrogen™ Supplier No. RP98046
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59703. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RNA polymerases transcribe nuclear genes for ribosomal RNA and are integral components of ribosomal biogenesis. RNA polymerase I (Pol I) is located in the nucleolus and transcribes class I genes, which code for large ribosomal RNA. RNA polymerase II (Pol II) synthesizes mRNA. RNA polymerase III (Pol III) transcribes class III genes, encoding a number of small ribosomal RNA molecules including tRNA and 5S rRNA. RPAP1 (RNA polymerase II associated protein 1) is a 1, 393 amino acid nuclear protein that is thought to connect Pol II to acetylated Histone H3 and regulators of protein complex formation. A member of the RPAP1 family, RPAP1 exists as three alternatively spliced isoforms that are encoded by a gene that maps to human chromosome 15q15.1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BWH6
Concentration 5.1 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 26015
Name Human RPAP1 (aa 101-215) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1190005L06Rik; A730023M06Rik; AU041788; AW107702; Kiaa1403; mKIAA1403; RNA polymerase II associated protein 1; RNA polymerase II-associated protein 1; RPAP1; RPAP-1
Common Name RPAP1
Gene Symbol RPAP1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HITAVLTKIIERDTSSVAVNLPVPSGVAFPAVFLRSRDTQGKSATSGKRSIFAQEIAARRIAEAKGPSVGEVVPNVGPPEGAVTCETPTPRNQGCQLPGSSHSFQGPNLVTGKGL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.