Learn More
Sino Biological Anti-RPL26 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109611T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human RPL26, and purified by antigen affinity chromatography.
Catalog No. 50-241-0782