Learn More
Sino Biological Human RPS10 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50136R076100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human RPS10 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4490