Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RPUSD3 (aa 83-163) Control Fragment Recombinant Protein

Catalog No. RP96384
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96384 Supplier Invitrogen™ Supplier No. RP96384
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58001. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RNA Pseudouridine Synthase D3 (RPUSD3) is a gene that encodes a pseudouridine synthase, an enzyme responsible for catalyzing the isomerization of uridine to pseudouridine in RNA molecules. This post-transcriptional modification is crucial for the stability and function of tRNAs and rRNAs. RPUSD3, along with other pseudouridine synthases such as PUS1, PUSL1, and TRUB2, is predicted to localize to mitochondria, indicating its role in mitochondrial RNA processing and maturation. The pseudouridylation activity provided by RPUSD3 is essential for the proper folding and functionality of mitochondrial RNAs, which are fundamental for the biogenesis of the mitochondrial electron transport chain and ATP synthase. While detailed structural insights into RPUSD3 are not yet fully characterized, its enzymatic function is critical in maintaining cellular energy metabolism and overall mitochondrial health.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P087
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 285367
Name Human RPUSD3 (aa 83-163) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI527266; Mitochondrial mRNA pseudouridine synthase RPUSD3; RGD1307582; RNA pseudouridylate synthase domain containing 3; RNA pseudouridylate synthase domain-containing protein 3; Rpusd3
Common Name RPUSD3
Gene Symbol RPUSD3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VVDRKGPLVTLNKPQGLPVTGKPGELTLFSVLPELSQSLGLREQELQVVRASGKESSGLVLLSSCPQTASRLQKYFTHARR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.